Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
E2F7, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of E2F7.
Supplier: Abnova Corporation PAB12815
Description
E2F transcription factors, such as E2F7, play an essential role in the regulation of cell cycle progression (Di Stefano et al., 2003 [PubMed 14633988]).[supplied by OMIM
Sequence: C-DLSKQKKFTPERNPSpecifications
E2F7 | |
Polyclonal | |
Unconjugated | |
ELISA (1:16000) Western Blot ( 0.1-0.3 ug/mL) The optimal working dilution should be determined by the end user. | |
E2F7 | |
FLJ12981 | |
Goat | |
Antigen affinity purification | |
RUO | |
144455 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Western Blot | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of E2F7. | |
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
NP_976328.2 | |
E2F7 | |
A synthetic peptide corresponding to internal region of human E2F7. | |
100 μg | |
Primary | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction