Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ELMO1, Goat, Polyclonal Antibody, Abnova™

Goat polyclonal antibody raised against synthetic peptide of ELMO1.

Supplier:  Abnova Corporation PAB6103

Catalog No. 89-132-147


Add to Cart

Description

Description

The protein encoded by this gene interacts with the dedicator of cyto-kinesis 1 protein to promote phagocytosis and effect cell shape changes. Similarity to a C. elegans protein suggests that this protein may function in apoptosis and in cell migration. Alternative splicing of this gene results in multiple transcript variants encoding different isoforms. [provided by RefSeq

Sequence: PKEPSNYDFVYDCN
Specifications

Specifications

ELMO1
Polyclonal
Unconjugated
ELISA (1:32000) Western blot (1-2 ug/mL) The optimal working dilution should be determined by the end user.
ELMO1
CED-12/CED12/ELMO-1/KIAA0281/MGC126406
Goat
Antigen affinity purification
RUO
9844
Bovine, Fish, Human
Liquid
ELISA, Immunoprecipitation, Western Blot
0.5 mg/mL
Goat polyclonal antibody raised against synthetic peptide of ELMO1.
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
NP_055615.8;NP_569709.1;NP_0010.34548.1
ELMO1
A synthetic peptide corresponding to C-terminus of human ELMO1.
100 μg
Primary
This antibody is expected to recognize both human isoforms of this protein. NP_569709.1 and NP_0010.34548.1 represent the same isoform.
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only