Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FANCL, Goat, Polyclonal Antibody, Abnova™

Goat polyclonal antibody raised against synthetic peptide of FANCL.

Manufacturer:  Abnova Corporation PAB19015

Catalog No. 89-120-568


Add to cart

Description

Description

The Fanconi anemia complementation group (FANC) currently includes FANCA, FANCB, FANCC, FANCD1 (also called BRCA2), FANCD2, FANCE, FANCF, FANCG, FANCI, FANCJ (also called BRIP1), FANCL, FANCM and FANCN (also called PALB2). The previously defined group FANCH is the same as FANCA. Fanconi anemia is a genetically heterogeneous recessive disorder characterized by cytogenetic instability, hypersensitivity to DNA crosslinking agents, increased chromosomal breakage, and defective DNA repair. The members of the Fanconi anemia complementation group do not share sequence similarity; they are related by their assembly into a common nuclear protein complex. This gene encodes the protein for complementation group L. Alternative splicing results in two transcript variants encoding different isoforms. [provided by RefSeq

Sequence: C-QNLKDVLEIDFP
Specifications

Specifications

FANCL
0.5 mg/mL
Goat polyclonal antibody raised against synthetic peptide of FANCL.
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
NP_001108108.1;NP_060532.2
FAAP43|FLJ10335|PHF9|POG
Goat
Antigen affinity purification
RUO
Primary
55120
ELISA, Western Blot
Unconjugated
ELISA (1:16000) Western Blot (1-3 ug/mL) The optimal working dilution should be determined by the end user.
FANCL
Liquid
FANCL
A synthetic peptide corresponding to amino acids at internal region of human FANCL.
100 ug
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only