Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FMR1 Goat anti-Human, Polyclonal Antibody, Abnova™

Goat polyclonal antibody raised against synthetic peptide of FMR1.

Manufacturer:  Abnova Corporation PAB14144

Catalog No. 89-116-957


Add to cart

Description

Description

The protein encoded by this gene binds RNA and is associated with polysomes. The encoded protein may be involved in mRNA trafficking from the nucleus to the cytoplasm. A trinucleotide repeat (CGG) in the 5' UTR is normally found at 6-53 copies, but an expansion to 55-230 repeats is the cause of fragile X syndrome. Expansion of the trinucleotide repeat may also cause one form of premature ovarian failure (POF1). [provided by RefSeq

Sequence: C-NPNKPATKDTFHKIK
Specifications

Specifications

FMR1
0.5 mg/mL
Goat polyclonal antibody raised against synthetic peptide of FMR1.
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
NP_002015.1
FMRP|FRAXA|MGC87458|POF|POF1
Goat
Antigen affinity purification
RUO
Primary
2332
Human
ELISA, Western Blot
Unconjugated
ELISA (1:32000) Western Blot (0.3-1 ug/mL) The optimal working dilution should be determined by the end user.
FMR1
Liquid
FMR1
A synthetic peptide corresponding to amino acids 116-130 of human FMR1.
100 ug
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Approx 70 Da band observed in lysates of cell lines HeLa and Jurkat (calculated MW of 71.2 KDa according to NP_002015.1).
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only