Learn More
FOXA1, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of FOXA1.
Supplier: Abnova Corporation PAB6418
Description
This gene encodes a member of the forkhead class of DNA-binding proteins. These hepatocyte nuclear factors are transcriptional activators for liver-specific transcripts such as albumin and transthyretin, and they also interact with chromatin. Similar family members in mice have roles in the regulation of metabolism and in the differentiation of the pancreas and liver. [provided by RefSeq
Sequence: C-GVYSRPVLNTSSpecifications
FOXA1 | |
Polyclonal | |
Unconjugated | |
ELISA (1:16000) Western blot (0.2-0.6 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5-10 ug/mL) The optimal working dilution should be determined by the end user. | |
FOXA1 | |
HNF3A/MGC33105/TCF3A | |
Goat | |
Antigen affinity purification | |
RUO | |
3169 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ChIP Assay, ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of FOXA1. | |
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
NP_004487.2 | |
FOXA1 | |
A synthetic peptide corresponding to C-terminus of human FOXA1. | |
100 μg | |
Primary | |
Human, Mouse | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only