Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FOXL1, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of FOXL1.
Supplier: Abnova Corporation PAB6436
Description
Sequence: C-YFPLQVPDTVLHFQSpecifications
FOXL1 | |
Polyclonal | |
Unconjugated | |
ELISA (1:16000) The optimal working dilution should be determined by the end user. | |
FOXL1 | |
FKH6/FKHL11/FREAC7 | |
Goat | |
Antigen affinity purification | |
RUO | |
2300 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of FOXL1. | |
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
NP_005241.1 | |
FOXL1 | |
A synthetic peptide corresponding to human FOXL1. | |
100 μg | |
Primary | |
Human | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only