Learn More
FYN, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of FYN.
Supplier: Abnova Corporation PAB6050
Description
This gene is a member of the protein-tyrosine kinase oncogene family. It encodes a membrane-associated tyrosine kinase that has been implicated in the control of cell growth. The protein associates with the p85 subunit of phosphatidylinositol 3-kinase and interacts with the fyn-binding protein. Alternatively spliced transcript variants encoding distinct isoforms exist. [provided by RefSeq
Sequence: VQCKDKEATKLTESpecifications
FYN | |
Polyclonal | |
Unconjugated | |
ELISA (1:32000) Western Blot (0.2-1 ug/mL) The optimal working dilution should be determined by the end user. | |
FYN | |
MGC45350/SLK/SYN | |
Goat | |
Antigen affinity purification | |
RUO | |
2534 | |
Human, Mouse | |
Liquid |
ELISA, Western Blot | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of FYN. | |
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
NP_002028.1;NP_694592.1;NP_694593.1 | |
FYN | |
A synthetic peptide corresponding to human FYN. | |
100 μg | |
Primary | |
This antibody is expected to recognize all three reported isoforms (as represented by NP_002028.1, NP_694592.1, NP_694593.1). | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only