Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPR3, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of GPR3.
Supplier: Abnova Corporation PAB7560
Description
Sequence: C-LQRHLLPASHSpecifications
GPR3 | |
Polyclonal | |
Unconjugated | |
ELISA (1:32000) Western Blot (0.3-1 ug/mL) The optimal working dilution should be determined by the end user. | |
GPR3 | |
ACCA | |
Goat | |
Antigen affinity purification | |
RUO | |
2827 | |
Human | |
Liquid |
ELISA, Western Blot | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of GPR3. | |
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
NP_005272.1 | |
GPR3 | |
A synthetic peptide corresponding to human GPR3. | |
100 μg | |
Primary | |
This antibody is not expected to cross-react with GPR6 and GPR12. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only