Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPX6, Goat, Polyclonal Antibody, Abnova™

Goat polyclonal antibody raised against synthetic peptide of GPX6.

Supplier:  Abnova Corporation PAB7066

Catalog No. 89-132-922


Add to Cart

Description

Description

This gene product belongs to the glutathione peroxidase family, which functions in the detoxification of hydrogen peroxide. It contains a selenocysteine (Sec) residue at its active site. The selenocysteine is encoded by the UGA codon, which normally signals translation termination. The 3' UTR of Sec-containing genes have a common stem-loop structure, the sec insertion sequence (SECIS), which is necessary for the recognition of UGA as a Sec codon rather than as a stop signal. Expression of this gene is restricted to embryos and adult olfactory epithelium. [provided by RefSeq

Sequence: QTLKPQNRKVDC
Specifications

Specifications

GPX6
Polyclonal
Unconjugated
ELISA (1:64000) The optimal working dilution should be determined by the end user.
GPX6
GPX6
A synthetic peptide corresponding to human GPX6.
100 μg
Primary
Human
Liquid
ELISA
0.5 mg/mL
Goat polyclonal antibody raised against synthetic peptide of GPX6.
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
NP_874360.1
Goat
Antigen affinity purification
RUO
257202
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only