Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HADH, Goat, Polyclonal Antibody, Abnova™

Goat polyclonal antibody raised against synthetic peptide of HADH.

Supplier:  Abnova Corporation PAB7412

Catalog No. 89-133-230


Add to Cart

Description

Description

This gene is a member of the 3-hydroxyacyl-CoA dehydrogenase gene family. The encoded protein functions in the mitochondrial matrix to catalyze the oxidation of straight-chain 3-hydroxyacyl-CoAs as part of the beta-oxidation pathway. Its enzymatic activity is highest with medium-chain-length fatty acids. Mutations in this gene cause one form of familial hyperinsulinemic hypoglycemia. The human genome contains a related pseudogene. [provided by RefSeq

Sequence: C-YERGDASKEDID
Specifications

Specifications

HADH
Polyclonal
Unconjugated
ELISA (1:64000) Western Blot (0.01-0.03 ug/mL) The optimal working dilution should be determined by the end user.
HADH
HADH
A synthetic peptide corresponding to human HADH.
100 μg
Primary
Human
Liquid
ELISA, Western Blot
0.5 mg/mL
Goat polyclonal antibody raised against synthetic peptide of HADH.
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
HAD/HADH1/HADHSC/HHF4/M/SCHAD/MGC8392/SCHAD
Goat
Antigen affinity purification
RUO
3033
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only