Learn More
HOXA9 Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of HOXA9.
Supplier: Abnova Corporation PAB18560
Description
In vertebrates, the genes encoding the class of transcription factors called homeobox genes are found in clusters named A, B, C, and D on four separate chromosomes. Expression of these proteins is spatially and temporally regulated during embryonic development. This gene is part of the A cluster on chromosome 7 and encodes a DNA-binding transcription factor which may regulate gene expression, morphogenesis, and differentiation. This gene is highly similar to the abdominal-B (Abd-B) gene of Drosophila. A specific translocation event which causes a fusion between this gene and the NUP98 gene has been associated with myeloid leukemogenesis. [provided by RefSeq
Sequence: C-QPPRQAATLAEHPDSpecifications
HOXA9 | |
Polyclonal | |
Unconjugated | |
ELISA (1:16000) The optimal working dilution should be determined by the end user. | |
HOXA9 | |
HOXA9 | |
A synthetic peptide corresponding to amino acids 37-50 at internal region of human HOXA9. | |
100 μg | |
Primary | |
Human | |
Liquid |
ELISA | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of HOXA9. | |
In 0.5mg/mL Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA) | |
ABD-B/HOX1/HOX1.7/HOX1G/MGC1934 | |
Goat | |
Antigen affinity purification | |
RUO | |
3205 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only