Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HPGD, Goat, Polyclonal Antibody, Abnova™

Goat polyclonal antibody raised against synthetic peptide of HPGD.

Supplier:  Abnova Corporation PAB6712

Catalog No. 89-132-621


Add to Cart

Description

Description

This gene encodes a member of the short-chain nonmetalloenzyme alcohol dehydrogenase protein family. The encoded enzyme is responsible for the metabolism of prostaglandins, which function in a variety of physiologic and cellular processes such as inflammation. Mutations in this gene result in primary autosomal recessive hypertrophic osteoarthropathy and cranioosteoarthropathy. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Sequence: C-DYDTTPFQAKTQ
Specifications

Specifications

HPGD
Polyclonal
Unconjugated
ELISA (1:128000) Western blot (0.1-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (2-4 ug/mL) The optimal working dilution should be determined by the end user.
HPGD
HPGD
A synthetic peptide corresponding to human HPGD.
100 μg
Primary
Human
Liquid
ELISA, Immunohistochemistry (PFA fixed), Western Blot
0.5 mg/mL
Goat polyclonal antibody raised against synthetic peptide of HPGD.
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
15-PGDH/PGDH/PGDH1/SDR36C1
Goat
Antigen affinity purification
RUO
3248
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only