Learn More
Goat anti-Human IgA (Heavy chain) Recombinant Secondary Antibody, HRP, Invitrogen™
Goat Recombinant Monoclonal Secondary Antibody
Specifications
Antigen | IgA (Heavy chain) |
---|---|
Clone | 43H46L44 |
Concentration | 1.0 mg/mL |
Applications | ELISA, Western Blot |
Classification | Recombinant Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
A55737
![]() |
LIFE TECHNOLOGIES
A55737 |
250 μL | This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
A5573750UL
![]() |
LIFE TECHNOLOGIES
A5573750UL |
50 μL | This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
This Goat anti-Human IgA (Heavy chain), Recombinant Monoclonal Antibody specifically detects human IgA heavy chain with no cross-reactivity to human IgG or IgM or various species immunoglobulins. The sensitivity and specificity of each lot is confirmed using ELISA. Recombinant antibodies are produced using specific genes that code for the desired antibodies. These genes are cloned into an expression vector and expressed in vitro. The advantages of recombinant antibodies include: better specificity, animal origin-free formulation, and lot-to-lot consistency. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
Human IgA (Heavy chain) Recombinant Monoclonal antibody specifically detects Human IgA (Heavy chain) in Human samples. It is validated for ELISA, Western BlotSpecifications
IgA (Heavy chain) | |
1.0 mg/mL | |
Recombinant Monoclonal | |
Liquid | |
RUO | |
PBS with proprietary stabilizer and proprietary preservative; pH 7.4 | |
IgG | |
Protein G | |
Secondary Antibody |
43H46L44 | |
ELISA, Western Blot | |
HRP | |
Goat | |
Human | |
Purified human IgA | |
Secondary | |
4° C, do not freeze |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.