Learn More
IgE Fc Goat anti-Human, Invitrogen™ Non-distribution product as customer accommodation.
Goat Polyclonal Secondary Antibody
Manufacturer:  Invitrogen H15700
Catalog No. 50-112-1283
Add to cart
Description
Description
Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
Specifications
Goat anti-Human IgE Fc Secondary Antibody
Polyclonal
PBS with 0.1% sodium azide
Goat
Purified
ASR
Secondary Antibody
Polyclonal
Flow Cytometry
Unconjugated
Liquid
IgG
1 mL
4° C, store in dark
Secondary
Human