Goat anti-Human IgG, Affinity Purified F(ab')2, Secondary Antibody Millipore™ (Chemicon™)

Manufacturer: EMD Millipore AQ112F/DEL

 View more versions of this product

Catalog No. 50-174-524

 N/A
N/A

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    Format Semi-purified
    Antigen Goat anti-Human IgG, Affinity Purified F(ab')2
    Host Species Goat
    Quantity 1mg
    Regulatory Status RUO
    Primary or Secondary Secondary
    Monoclonal or Polyclonal Polyclonal
    Conjugate FITC
    Cross Reactivity Human

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit