Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ig Fab Goat anti-Human, Unconjugated, Polyclonal, Southern Biotech™
Goat Polyclonal Secondary Antibody
Supplier: Southern Biotechnology Associates 208501
Description
SouthernBiotech is a leading provider of polyclonal secondary antibodies offering high sensitivity and low background for use in a variety of applications.
Specifications
Ig Fab | |
Polyclonal | |
Unconjugated | |
Goat | |
Affinity Purified | |
RUO | |
Human | |
IgG |
ELISA | |
0.5mg/mL | |
Supplied in borate buffered saline, pH 8.2; No preservatives or amine-containing buffer salts added | |
Human IgG, IgM, and IgA | |
0.5 mg | |
Secondary | |
Store at 2-8°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction