Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat anti-Human IgG Fc Secondary Antibody, Invitrogen™

Goat Polyclonal Secondary Antibody

Supplier:  Thermo Scientific PA185606

Catalog No. PA185606


Add to Cart

Description

Description

Goat anti Human IgG (Fc) antibody recognizes human IgG (Fc).Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-goat serum, human IgG, human IgG F(c)and human serum. Goat anti Human IgG (Fc) antibody does not recognize human IgG F(ab')2.

Human IgG Fc Polyclonal antibody specifically detects Human IgG Fc in Human samples. It is validated for ELISA, Immunoprecipitation, Western Blot.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Human IgG Fc
Polyclonal
Unconjugated
Goat
Affinity chromatography
RUO
Human
Liquid
ELISA, Immunoprecipitation, Western Blot
4 mg/mL
PBS with <0.1% sodium azide; pH 7.2
Human IgG (Fc) fragment.
2 mg
Secondary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
IgG
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit