Learn More
Goat anti-Human IgG1 (Heavy chain) Recombinant Secondary Antibody, Alexa Fluor™ 647, Invitrogen™
Goat Recombinant Monoclonal Secondary Antibody
$101.00
Specifications
Antigen | IgG1 (Heavy chain) |
---|---|
Clone | 3H60L9 |
Concentration | 1.0 mg/mL |
Applications | Immunocytochemistry |
Classification | Recombinant Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
A5574350UL
![]() |
LIFE TECHNOLOGIES
A5574350UL |
50 μL |
Each for $101.00
|
|
A55743
![]() |
LIFE TECHNOLOGIES
A55743 |
250 μL | This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
This Goat anti-Human IgG1 (Heavy chain) Recombinant Monoclonal Antibody specifically detects human IgG1 heavy chain with no cross-reactivity to human IgG2, IgG3, IgG4, IgA or IgM or various species immunoglobulins, including monkey IgG. The sensitivity and specificity of each lot is confirmed using ELISA. Recombinant antibodies are produced using specific genes that code for the desired antibodies. These genes are cloned into an expression vector and expressed in vitro. The advantages of recombinant antibodies include: better specificity, animal origin-free formulation, and lot-to-lot consistency. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
Human IgG1 (Heavy chain) Recombinant Monoclonal antibody specifically detects Human IgG1 (Heavy chain) in Human samples. It is validated for ImmunocytochemistrySpecifications
IgG1 (Heavy chain) | |
1.0 mg/mL | |
Recombinant Monoclonal | |
Liquid | |
RUO | |
PBS with 5mM sodium azide; pH 7.4 | |
IgG | |
Protein G | |
Secondary Antibody |
3H60L9 | |
Immunocytochemistry | |
Alexa Fluor 647 | |
Goat | |
Human | |
Purified Human IgG | |
Secondary | |
4° C, store in dark, DO NOT FREEZE! |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.