Learn More
Goat anti-Human Kappa Light Chain Secondary Antibody, Invitrogen™
Goat Polyclonal Secondary Antibody
Supplier: Thermo Scientific 31129
Description
Product # 31129 has been successfully used in Western blot, IF, ICC, ISH, ELISA and IHC applications. Product # 31129 reacts with the kappa light chains of human IgG, but does not react against non-immunoglobulin serum proteins. Based on solid-phase enzyme immunoassay, this antibody has minimal cross-reactivity with human gamma chain (1%), human lambda chain and human lambda IgG. However, this antibody may cross-react with immunoglobulins from other species. The activity measured by a solid phase binding assay is 97%.
Human Kappa Light Chain Polyclonal antibody specifically detects Human Kappa Light Chain in Human samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry, In Situ Hybridization (ISH), Western Blot.Specifications
Human Kappa Light Chain | |
Polyclonal | |
Unconjugated | |
Goat | |
500 μg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Glycerol (1:1) may be added for added stability. | |
IgG |
ELISA, Immunocytochemistry, Immunohistochemistry, In Situ Hybridization (ISH), Western Blot | |
1 mg/mL | |
PBS with 20mg/mL sucrose and 0.08% sodium azide; pH 7.8 | |
Antigen affinity chromatography | |
RUO | |
Human | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.