Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kappa Goat anti-Human, AP, Polyclonal, Southern Biotech™
Goat Polyclonal Secondary Antibody
Supplier: Southern Biotechnology Associates 206004
Description
SouthernBiotech is a leading provider of polyclonal secondary antibodies offering high sensitivity and low background for use in a variety of applications.
Specifications
Kappa | |
Polyclonal | |
Alkaline Phosphatase (AP) | |
Goat | |
Affinity Purified | |
RUO | |
Human | |
IgG |
ELISA, Dot Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
Lot specific | |
Supplied in a stock solution 50 mM Tris/1 mM MgCl2/50% glycerol, pH 8.0, containing NaN3 as preservative | |
Human κ light chains | |
1.0 mL | |
Secondary | |
Store at 2-8°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction