Learn More
Saccharomyces cerevisiae, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against Saccharomyces cerevisiae.
Manufacturer:  Abnova Corporation PAB13821
Catalog No. 89-137-693
Add to cart
Description
Description
Specifications
Specifications
Saccharomyces cerevisiae
Unconjugated
The optimal working dilution should be determined by the end user.
Liquid
Native from solubilized yeast cells.
RUO
Primary
Saccharomyces cerevisiae. Reacts with more than 40 proteins by Western blot.
Dot Blot, ELISA, Western Blot
Goat polyclonal antibody raised against Saccharomyces cerevisiae.
In antiserum
Goat
1 mL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Yeast