Learn More
IGF2BP1, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of IGF2BP1.
Supplier: Abnova Corporation PAB18605
Description
This gene encodes a member of the insulin-like growth factor 2 mRNA-binding protein family. The protein encoded by this gene contains four K homology domains and two RNA recognition motifs. It functions by binding to the mRNAs of certain genes, including insulin-like growth factor 2, beta-actin and beta-transducin repeat-containing protein, and regulating their translation. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: C-EKVFAEHKISYSGQSpecifications
IGF2BP1 | |
Polyclonal | |
Unconjugated | |
ELISA (1:16000) Western Blot (0.3-1 ug/mL) The optimal working dilution should be determined by the end user. | |
IGF2BP1 | |
IGF2BP1 | |
A synthetic peptide corresponding to amino acids at internal region of human IGF2BP1. | |
100 μg | |
Primary | |
This antibody is expected to recognize both reported isoforms (NP_006537.3; NP_001153895.1). | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Western Blot | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of IGF2BP1. | |
In 0.5mg/mL Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA) | |
CRD-BP/CRDBP/IMP-1/IMP1/VICKZ1/ZBP1 | |
Goat | |
Antigen affinity purification | |
RUO | |
10642 | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only