Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL25 Goat anti-Human, Polyclonal Antibody, Abnova™

Goat polyclonal antibody raised against synthetic peptide of IL25.

Supplier:  Abnova Corporation PAB27686

Catalog No. 89-129-201


Add to Cart

Description

Description

The protein encoded by this gene is a cytokine that shares the sequence similarity with IL17. This cytokine can induce NF-kappaB activation, and stimulate the production of IL8. Both this cytokine and IL17B are ligands for the cytokine receptor IL17BR. Studies of the similar gene in mice suggested that this cytokine may be a proinflammatory cytokine favoring Th2-type immune response. Two alternatively spliced transcript variants of this gene encoding distinct isoforms have been reported. [provided by RefSeq

Sequence: RPNRHPESCRASED
Specifications

Specifications

IL25
Polyclonal
Unconjugated
ELISA (1:128000) Western Blot (0.3-1ug/ml) The optimal working dilution should be determined by the end user.
IL25
IL25
A synthetic peptide corresponding to amino acids 66-79 at internal region of human IL25.
100 μg
Primary
Human
Western Blot
0.5 mg/mL
Goat polyclonal antibody raised against synthetic peptide of IL25.
In Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA)
IL-17E/IL-25/IL17E
Goat
Affinity Purified
RUO
64806
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only