Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LMX1B, Goat, Polyclonal Antibody, Abnova™

Goat polyclonal antibody raised aganist synthetic peptide of LMX1B.

Manufacturer:  Abnova Corporation PAB15644

Catalog No. 89-117-712


Add to cart

Description

Description

Sequence: QQALTTSCYFRDRK
Specifications

Specifications

LMX1B
0.5 mg/mL
Goat polyclonal antibody raised aganist synthetic peptide of LMX1B.
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Liquid
LMX1B
A synthetic peptide corresponding to amino acids at internal region of human LMX1B.
100 ug
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
ELISA
Unconjugated
ELISA (1:64000) The optimal working dilution should be determined by the end user.
LMX1B
LMX1.2|MGC138325|MGC142051|NPS1
Goat
Antigen affinity purification
RUO
Primary
4010
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only