Learn More
LSM2, Goat, Polyclonal Antibody, Abnova
Goat polyclonal antibody raised against synthetic peptide of LSM2.
Manufacturer: Abnova Corporation PAB25458
Description
Sm-like proteins were identified in a variety of organisms based on sequence homology with the Sm protein family (see SNRPD2; MIM 601061). Sm-like proteins contain the Sm sequence motif, which consists of 2 regions separated by a linker of variable length that folds as a loop. The Sm-like proteins are thought to form a stable heteromer present in tri-snRNP particles, which are important for pre-mRNA splicing.[supplied by OMIM
Sequence: CDQYLNIKLTDISVTSpecifications
LSM2 | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of LSM2. | |
In Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA) | |
Liquid | |
LSM2 | |
A synthetic peptide corresponding to amino acids 33-46 at internal region of human LSM2. | |
100 ug | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
Polyclonal | |
Human |
ELISA, Western Blot | |
Unconjugated | |
ELISA (1:2000) Western Blot (1-3 ug/mL) The optimal working dilution should be determined by the end user. | |
LSM2 | |
C6orf28|G7b|YBL026W|snRNP | |
Goat | |
Antigen affinity purification | |
RUO | |
Primary | |
57819 |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only