Learn More
malondialdehyde, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against malondialdehyde.
Manufacturer:  Abnova Corporation PAB1288
Catalog No. 89-137-558
Add to cart
Description
Description
Specifications
Specifications
malondialdehyde
Unconjugated
Western Blot (1:4000-1:20000) ELISA (1:4000-1:20000) The optimal working dilution should be determined by the end user.
Liquid
Malondialdehyde conjugated with KLH.
RUO
Primary
This antibody specifically binds to MDA-LDL and other MDA modified proteins.
ELISA, Western Blot
Goat polyclonal antibody raised against malondialdehyde.
In 75mM PBS, 75mM NaCl, 0.5mM EDTA, pH 7.2 (0.02% sodium azide)
Goat
500 ug
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal