Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MARK2, Goat, Polyclonal Antibody, Abnova™

Goat polyclonal antibody raised against synthetic peptide of MARK2.

Supplier:  Abnova Corporation PAB7578

Catalog No. 89-133-383


Add to Cart

Description

Description

This gene encodes a member of the Par-1 family of serine/threonine protein kinases. The protein is an important regulator of cell polarity in epithelial and neuronal cells, and also controls the stability of microtubules through phosphorylation and inactivation of several microtubule-associating proteins. The protein localizes to cell membranes. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Sequence: C-QNGKDSTAPQR
Specifications

Specifications

MARK2
Polyclonal
Unconjugated
ELISA (1:16000) Western Blot (0.3-1 ug/mL) The optimal working dilution should be determined by the end user.
MARK2
MARK2
A synthetic peptide corresponding to human MARK2.
100 μg
Primary
This antibody is to recognize the reported isoforms (NP_001034557.1; NP_004945.3).
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
ELISA, Western Blot
0.5 mg/mL
Goat polyclonal antibody raised against synthetic peptide of MARK2.
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
EMK1/MGC99619/PAR-1/Par1b
Goat
Antigen affinity purification
RUO
2011
Human
Liquid
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only