Learn More
MAX, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of MAX.
Supplier: Abnova Corporation PAB6023
Description
The protein encoded by this gene is a member of the basic helix-loop-helix leucine zipper (bHLHZ) family of transcription factors. It is able to form homodimers and heterodimers with other family members, which include Mad, Mxi1 and Myc. Myc is an oncoprotein implicated in cell proliferation, differentiation and apoptosis. The homodimers and heterodimers compete for a common DNA target site (the E box) and rearrangement among these dimer forms provides a complex system of transcriptional regulation. Multiple alternatively spliced transcript variants have been described for this gene but the full-length nature for some of them is unknown. [provided by RefSeq
Sequence: C-EEPQSRKKLRMEASSpecifications
MAX | |
Polyclonal | |
Unconjugated | |
ELISA (1:32000) Western blot (0.1-0.3 ug/mL) The optimal working dilution should be determined by the end user. | |
MAX | |
MAX | |
A synthetic peptide corresponding to human MAX. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Liquid |
ELISA, Western Blot | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of MAX. | |
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
MGC10775/MGC11225/MGC18164/MGC34679/MGC36767/bHLHd4/bHLHd5/bHLHd6/bHLHd7/bHLHd8/orf1 | |
Goat | |
Antigen affinity purification | |
RUO | |
4149 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only