Learn More
Goat anti-Monkey IgM mu heavy chain, Polyclonal, Secondary Antibody, Abnova™
Manufacturer:  Abnova Corporation PAB9327
Catalog No. 89-138-997
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Specifications
Specifications
Goat anti-Monkey IgM mu heavy chain
Unconjugated
The optimal working dilution should be determined by the end user.
Liquid
Monkey IgM mu heavy chain
1 mg
Secondary
No reaction was observed against monkey IgG or monkey IgA. Specificity was confirmed by ELISA at less than 1% cross reactivity against other monkey heavy or light chain isotypes.
ELISA, Immunohistochemistry, Western Blot
Goat anti-monkey IgG recognizes monkey IgM mu heavy chain. This secondary antibody was purified using antigen affinity chromatography.
In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide)
Goat
This product was prepared from monospecific antiserum by immunoaffinity chromatography, followed by solid phase adsorption(s) to remove any unwanted reactivities.
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Monkey