Learn More
Goat anti-Mouse IgG - H&L , Secondary Antibody, Abcam™
This product is for research use only and not for diagnostic or therapeutic procedures. Customer is granted a limited, non-exclusive license for internal research and shall not attempt to reverse engineer.
Supplier: Abcam AB67851MG

Description
Host Species: Goat
Isotype: IgG
Immunogen: mouse whole molecule
Applications:
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunomicroscopy, Western Blotting
Specifications
Mouse IgG- H&L | |
2mg/mL | |
Goat | |
Immunology | |
IgG |
Polyclonal | |
FITC | |
1 mg | |
Secondary |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.