Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Goat anti-Mouse IgG, H+L, Secondary Antibody ™ (Chemicon™)

Supplier:  MilliporeSigma™ AP124B/DEL

Catalog No. 50-174-394


Description

Description

Isotype: IgG
Host Species: Goat
Cross Reactivity: Mouse
Format: Immunoaffinity Purified

ELISA, Immunoelectrophoresis, Immunoprecipitation

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Goat anti-Mouse IgG, H+L
Biotin
Affinity Purified
RUO
Secondary
Polyclonal
Goat
2 mL
Enzyme
Mouse
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.