Learn More
IgG (H+L) Cross-Adsorbed Goat anti-Mouse, PE-Alexa Fluor® 647, Invitrogen™ Non-distribution product as customer accommodation.
Goat Polyclonal Secondary Antibody
Manufacturer:  Invitrogen A20990
Catalog No. 50-113-7342
Add to cart
Description
Description
Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
Specifications
Mouse IgG (H+L) Cross-Adsorbed
Polyclonal
PE-Alexa Fluor® 647
Liquid
Gamma Immunoglobins Heavy and Light chains
Purified
RUO
Secondary Antibody
Polyclonal
Flow Cytometry
1 mg/mL
PBS with 1% glycerol, 2mM EDTA and 5mM sodium azide; pH 7.5
Goat
IgG
100 μg
4° C, store in dark
Secondary
Mouse