Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Goat anti-Mouse IgG, Whole Molecule Polyclonal Secondary Antibody, MP Biomedicals
Specifications
Antigen | IgG, Whole Molecule |
---|---|
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Lyophilized |
Host Species | Goat |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
ICN55455
|
MP Biomedicals, Inc
0855455 |
5 mL |
Each of 1 for $265.00
|
N/A |
Description
Host Species: Goat
Isotype: IgG
Species Reactivity: Mouse
Specifications
IgG, Whole Molecule | |
Unconjugated | |
Goat | |
IgG |
Polyclonal | |
Lyophilized | |
Mouse | |
Secondary |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title