Learn More
Enzo Life SciencesSupplier Diversity Partner Goat anti-mouse IgM, polyclonal antibody (HRP conjugate) (1 ml)  DFS Item
Manufacturer:  Enzo Life SciencesSupplier Diversity Partner ADISAB110J
Host: Goat. Immunogen: Purified mouse IgM. Species Reactivity: Mouse. Specificity: Based on immunoelectrophoresis and/or ELISA, the antibody reacts with the heavy chain of mouse IgM but not with mouse IgG or the light chains of mouse immunoglobulins. No antibody was detected against non-immunoglobulin serum proteins. Crossreactivity: May cross-react with IgM from other species. Applications: ELISA, IHC, WB. Recommended dilutions: ELISA (1:2,500-50,000). Immunohistochemistry (1:250-2500). Western Blot (1:2500-50,000 colorimetric, 1:5,000-100,000 ECL). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Immunoaffinity purified. Formulation: Liquid. In 0.005M sodium phosphate, pH 7.6, containing 0.125M sodium chloride, 7.5mg/mL BSA, and 50% glycerol.
Catalog No. 50-667-385