Learn More
MTR, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of MTR.
Supplier: Abnova Corporation PAB6022
Description
MTR encodes the enzyme 5-methyltetrahydrofolate-homocysteine methyltransferase. This enzyme, also known as cobalamin-dependent methionine synthase, catalyzes the final step in methionine biosynthesis. Mutations in MTR have been identified as the underlying cause of methylcobalamin deficiency complementation group G. [provided by RefSeq
Sequence: C-VEKWLGPILGYDTDSpecifications
MTR | |
Polyclonal | |
Unconjugated | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections)(3-5 ug/mL) ELISA (1:32000) Western blot (1-3 ug/mL) The optimal working dilution should be determined by the end user. | |
MTR | |
MTR | |
A synthetic peptide corresponding to human MTR. | |
100 μg | |
Primary | |
Human | |
Liquid |
ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of MTR. | |
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
FLJ33168/FLJ43216/FLJ45386/MS | |
Goat | |
Antigen affinity purification | |
RUO | |
4548 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only