Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat anti-Muscle Phosphofructokinase/PFKM/PFK-1, Polyclonal, Novus Biologicals™

Goat Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP244228

Catalog No. NBP244228


Add to cart

Description

Description

Specifically detects Muscle Phosphofructokinase/PFKM/PFK-1 in Rabbit, Rat samples, and it is validated for ELISA, Immunocytochemistry, Immunofluorescence, Western Blotting

Muscle Phosphofructokinase/PFKM/PFK-1 Polyclonal antibody specifically detects Muscle Phosphofructokinase/PFKM/PFK-1 in Rat, Rabbit samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

Muscle Phosphofructokinase/PFKM/PFK-1
LYOPH
Western Blot 1:500 - 1:2000, ELISA 1:1000 - 1:5000, Immunocytochemistry/Immunofluorescence 1:200 - 1:2000
Purified
PFKM
Full length native Fructose-6-Phosphate Kinase purified from rabbit muscle. Uniprot ID P00511
IgG purified
RUO
Store lyophilized antibody at 4°C. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Polyclonal
Reconstitute with 100 ul deionized water.
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Unconjugated
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
6-phosphofructokinase, muscle type, EC 2.7.1,6-phosphofructo-1-kinase, EC 2.7.1.11, GSD7, MGC8699, PFK1, PFK-1, PFKA, PFK-A, PFKX, Phosphofructo-1-kinase isozyme A, Phosphofructokinase 1, phosphofructokinase, muscle, phosphofructokinase, polypeptide X, phosphofructokinase-M, phosphohexokinase
Goat
IgG
0.1 mg
Lipid and Metabolism
Primary
5213
Rat, Rabbit
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only