Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NEK7 Goat anti-Human, Mouse, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of NEK7.
Supplier: Abnova Corporation PAB18986
Description
NIMA-related kinases share high amino acid sequence identity with the gene product of the Aspergillus nidulans 'never in mitosis A' gene, which controls initiation of mitosis.[supplied by OMIM
Sequence: CINPDPEKRPDVTSpecifications
NEK7 | |
Polyclonal | |
Unconjugated | |
ELISA (1:32000) Western Blot (0.1-0.3 ug/mL) The optimal working dilution should be determined by the end user. | |
NEK7 | |
Goat | |
Antigen affinity purification | |
RUO | |
140609 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Western Blot | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of NEK7. | |
In 0.5mg/mL in Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
NEK7 | |
A synthetic peptide corresponding to amino acids at C-terminus of human NEK7. | |
100 μg | |
Primary | |
Human, Mouse | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction