Learn More
Influenza B, Goat, Biotin, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against yamagata strain of Influenza B.
Manufacturer:  Abnova Corporation PAB14282
Catalog No. 89-137-912
Add to cart
Description
Description
Specifications
Specifications
nfluenza B
Biotin
The optimal working dilution should be determined by the end user.
Liquid
Influenza B, strain Yamagata.
RUO
Primary
Purified virions. Does not cross react with influenza A virus, Parainfluenza virus (types 1-3), Respiratory Syncytial virus or Adenovirus. Negative vs monkey kidney cells by indirect immunofluorescence.
ELISA, Immunofluorescence, Western Blot
Goat polyclonal antibody raised against yamagata strain of Influenza B.
In 10mM PBS, pH 7.2 (0.09% sodium azide)
Goat
1 mL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Influenza virus