Learn More
NLGN3, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of NLGN3.
Supplier: Abnova Corporation PAB7418
Description
This gene encodes a member of a family of neuronal cell surface proteins. Members of this family may act as splice site-specific ligands for beta-neurexins and may be involved in the formation and remodeling of central nervous system synapses. Mutations in this gene may be associated with autism and Asperger syndrome. Multiple transcript variants encoding distinct isoforms have been identified for this gene, but their full length sequences have not been determined. [provided by RefSeq
Sequence: C-RKDKRRQEPLRQPSSpecifications
NLGN3 | |
Polyclonal | |
Unconjugated | |
ELISA (1:16000) Western Blot (0.05-0.2 ug/mL) The optimal working dilution should be determined by the end user. | |
NLGN3 | |
NLGN3 | |
A synthetic peptide corresponding to human NLGN3. | |
100 μg | |
Primary | |
Human | |
Liquid |
ELISA, Western Blot | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of NLGN3. | |
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
ASPGX1/AUTSX1/HNL3/KIAA1480 | |
Goat | |
Antigen affinity purification | |
RUO | |
54413 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only