Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NR2F6, Goat, Polyclonal Antibody, Abnova™

Goat polyclonal antibody raised against synthetic peptide of NR2F6.

Supplier:  Abnova Corporation PAB6246

Catalog No. 89-132-272

Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.


Description

Description

Sequence: C-STFNWPYGSGQ
Specifications

Specifications

NR2F6
Polyclonal
Unconjugated
ELISA (1:32000) The optimal working dilution should be determined by the end user.
NR2F6
NR2F6
A synthetic peptide corresponding to human NR2F6.
100 μg
Primary
Human
Liquid
ELISA
0.5 mg/mL
Goat polyclonal antibody raised against synthetic peptide of NR2F6.
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
EAR-2/EAR2/ERBAL2
Goat
Antigen affinity purification
RUO
2063
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only