Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OSCAR, Goat, Polyclonal Antibody, Abnova™

Goat polyclonal antibody raised against synthetic peptide of OSCAR.

Supplier:  Abnova Corporation PAB6984

Catalog No. 89-132-850


Add to Cart

Description

Description

Osteoclasts are multinucleated cells that resorb bone and are essential for bone homeostasis. This gene encodes an osteoclast-associated receptor (OSCAR), which is a member of the leukocyte receptor complex (LRC) protein family that plays critical roles in the regulation of both innate and adaptive immune responses. Different from the other LRC members, OSCAR expression is detected specifically in preosteoclasts or mature osteoclasts. OSCAR may be an important bone-specific regulator of osteoclast differentiation. Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Sequence: C-EGPEARPASSAP
Specifications

Specifications

OSCAR
Polyclonal
Unconjugated
ELISA (1:2000) The optimal working dilution should be determined by the end user.
OSCAR
OSCAR
A synthetic peptide corresponding to human OSCAR.
100 μg
Primary
This antibody is expected to recognize isoform 1 (NP_996554.1).
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
ELISA
0.5 mg/mL
Goat polyclonal antibody raised against synthetic peptide of OSCAR.
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
MGC33613/PIGR3
Goat
Antigen affinity purification
RUO
126014
Human
Liquid
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only