Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCBP4, Goat, Polyclonal Antibody, Abnova™

Goat polyclonal antibody raised against synthetic peptide of PCBP4.

Supplier:  Abnova Corporation PAB7325

Catalog No. 89-133-148


Add to Cart

Description

Description

This gene encodes a member of the KH-domain protein subfamily. Proteins of this subfamily, also referred to as alpha-CPs, bind to RNA with a specificity for C-rich pyrimidine regions. Alpha-CPs play important roles in post-transcriptional activities and have different cellular distributions. This gene is induced by the p53 tumor suppressor, and the encoded protein can suppress cell proliferation by inducing apoptosis and cell cycle arrest in G(2)-M. This gene's protein is found in the cytoplasm, yet it lacks the nuclear localization signals found in other subfamily members. Multiple alternatively spliced transcript variants have been described, but the full-length nature for only some has been determined. [provided by RefSeq

Sequence: C-NGSKKAERQKFSPY
Specifications

Specifications

PCBP4
Polyclonal
Unconjugated
ELISA (1:128000) Western Blot (1∼3 ug/mL) The optimal working dilution should be determined by the end user.
PCBP4
PCBP4
A synthetic peptide corresponding to human PCBP4.
100 μg
Primary
This antibody is expected to recognize all reported isoforms (NP_065151.1, NP_127501.1, NP_127502.1 and NP_12750.3.1).
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
ELISA, Western Blot
0.5 mg/mL
Goat polyclonal antibody raised against synthetic peptide of PCBP4.
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
LIP4/MCG10
Goat
Antigen affinity purification
RUO
57060
Human
Liquid
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only