Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PEBP4, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of PEBP4.
Manufacturer: Abnova Corporation PAB17819
Description
The phosphatidylethanolamine (PE)-binding proteins, including PEBP4, are an evolutionarily conserved family of proteins with pivotal biologic functions, such as lipid binding and inhibition of serine proteases (Wang et al., 2004 [PubMed 15302887]).[supplied by OMIM
Sequence: C-RERASEPKHKNQAESpecifications
PEBP4 | |
Polyclonal | |
Unconjugated | |
ELISA (1:4000) Western Blot (1-3 ug/mL) The optimal working dilution should be determined by the end user. | |
PEBP4 | |
PEBP4 | |
A synthetic peptide corresponding to human PEBP4. | |
100 μg | |
Primary | |
Human |
ELISA, Western Blot | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of PEBP4. | |
In 0.5mg/mL Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA) | |
CORK-1/CORK1/GWTM1933/MGC22776/PRO4408 | |
Goat | |
Antigen affinity purification | |
RUO | |
157310 | |
Liquid |
Safety and Handling
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only