Learn More
Goat anti-Pepsin, Polyclonal, Novus Biologicals™
Goat Polyclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP244230
Catalog No. NBP244230
Add to cart
Description
Description
Specifically detects Pepsin in Porcine samples, and it is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blotting
Pepsin Polyclonal antibody specifically detects Pepsin in Porcine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
Specifications
Pepsin
LYOPH
Western Blot 1:5000 - 1:20000, ELISA 1:50:000 - 1:200000, Immunohistochemistry 1:2000 - 1:10000, Immunoprecipitation 1:100, Immunohistochemistry-Paraffin
Purified
Goat
IgG
0.1 mg
Store lyophilized antibody at 4°C. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Polyclonal
Porcine
Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Unconjugated
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
DKFZp666J2410, ec 3.4.23.1, Pepsin a precursor, Pepsinogen 1 second locus, Pepsinogen 3 group 1, Pepsinogen 4 group I, Pepsinogen 5 group I, Pepsinogen A, Pepsinogen a3, Pepsinogen A4, Pepsinogen A5, Pepsinogen group 1, Pepsinogen I, Pga 3, Pga 4, Pga 5, pga3, pga4, pga5
Anti-Pepsin Antibody was produced by repeated immunizations with pork Pepsin protein isolated from porcine stomach.
IgG purified
RUO
Primary
Reconstitute with 100 ul deionized water.