Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIK3CA, Goat, Polyclonal Antibody, Abnova™

Goat polyclonal antibody raised against synthetic peptide of PIK3CA.

Supplier:  Abnova Corporation PAB7153

Catalog No. 89-132-997


Add to Cart

Description

Description

Phosphatidylinositol 3-kinase is composed of an 85kDa regulatory subunit and a 110kDa catalytic subunit. The protein encoded by this gene represents the catalytic subunit, which uses ATP to phosphorylate PtdIns, PtdIns4P and PtdIns(4,5)P2. This gene has been found to be oncogenic and has been implicated in cervical cancers. [provided by RefSeq

Sequence: C-HQWLKDKNKGEIYD
Specifications

Specifications

PIK3CA
Polyclonal
Unconjugated
ELISA (1:128000) Immunohistochemistry (2-3 ug/mL) The optimal working dilution should be determined by the end user.
PIK3CA
PIK3CA
A synthetic peptide corresponding to human PIK3CA.
100 μg
Primary
Human
Liquid
ELISA, Immunohistochemistry (PFA fixed)
0.5 mg/mL
Goat polyclonal antibody raised against synthetic peptide of PIK3CA.
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
MGC142161/MGC142163/PI3K/p110-alpha
Goat
Antigen affinity purification
RUO
5290
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only