Learn More
PLIN1 Goat anti-Human, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of PLIN1.
Supplier: Abnova Corporation PAB7250
Description
The protein encoded by this gene coats lipid storage droplets in adipocytes, thereby protecting them until they can be broken down by hormone-sensitive lipase. The encoded protein is the major cAMP-dependent protein kinase substrate in adipocytes and, when unphosphorylated, may play a role in the inhibition of lipolysis. Alternatively spliced transcript variants varying in the 5' UTR, but encoding the same protein, have been found for this gene. [provided by RefSeq]
Sequence: C-EPILGRAQYSQLRKKSpecifications
PLIN1 | |
Polyclonal | |
Unconjugated | |
ELISA (1:32000) Western Blot (0.01-0.03 ug/mL) Immunofluorescence (1 ug/mL) The optimal working dilution should be determined by the end user. | |
PLIN | |
PLIN | |
A synthetic peptide corresponding to human PLIN1. | |
100 μg | |
Primary | |
Human | |
Liquid |
ELISA, Immunofluorescence, Western Blot | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of PLIN1. | |
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
PERI | |
Goat | |
Antigen affinity purification | |
RUO | |
5346 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only