Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat anti-Porcine, IgM, Polyclonal, Novus Biologicals™

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP236559

Catalog No. NBP236559


Add to cart

Specifications

Specifications

IgM
1 mg/mL
Western Blot 1:1000-1:30000, ELISA 1:1000-1:30000, Immunohistochemistry 1:200-1:2000, Immunocytochemistry/Immunofluorescence 1:200-1:2000
Affinity Purified
Goat
IgG
1 ml
Store at 4C. Do not freeze.
Polyclonal
By immunoelectrophoresis and ELISA this antibody reacts specifically with pig IgM. This antibody may cross react with IgM from other species.
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Unconjugated
Phosphate Buffered Saline (PBS) with 0.09% Sodium Azide
AGM1, DKFZp686I15196, DKFZp686I15212, FLJ00385, immunoglobulin heavy constant mu, MGC104996, MGC52291, MU, VH
Pig IgM
Immunogen affinity purified
RUO
Secondary
3507
Porcine
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit