Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRDM16, Goat, Polyclonal Antibody, Abnova™

Goat polyclonal antibody raised against synthetic peptide of PRDM16.

Supplier:  Abnova Corporation PAB6224

Catalog No. 89-132-252


Add to Cart

Description

Description

The reciprocal translocation t(1;3)(p36;q21) occurs in a subset of myelodysplastic syndrome (MDS) and acute myeloid leukemia (AML). This gene is located near the 1p36.3 breakpoint and has been shown to be specifically expressed in the t(1:3)(p36,q21)-positive MDS/AML. The protein encoded by this gene is a zinc finger transcription factor and contains an N-terminal PR domain. The translocation results in the overexpression of a truncated version of this protein that lacks the PR domain, which may play an important role in the pathogenesis of MDS and AML. Alternatively spliced transcript variants encoding distinct isoforms have been reported. [provided by RefSeq

Sequence: C-TSESGAFHPINHL
Specifications

Specifications

PRDM16
Polyclonal
Unconjugated
ELISA (1:32000) The optimal working dilution should be determined by the end user.
PRDM16
PRDM16
A synthetic peptide corresponding to human PRDM16.
100 μg
Primary
Human
Liquid
ELISA
0.5 mg/mL
Goat polyclonal antibody raised against synthetic peptide of PRDM16.
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
KIAA1675/MEL1/MGC166915/PFM13
Goat
Antigen affinity purification
RUO
63976
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only