Learn More
PRDX2, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of PRDX2.
Supplier: Abnova Corporation PAB7050
Description
This gene encodes a member of the peroxiredoxin family of antioxidant enzymes, which reduce hydrogen peroxide and alkyl hydroperoxides. The encoded protein may play an antioxidant protective role in cells, and may contribute to the antiviral activity of CD8(+) T-cells. This protein may have a proliferative effect and play a role in cancer development or progression. The crystal structure of this protein has been resolved to 2.7 angstroms. Transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq
Sequence: C-NVDDSKEYFSKHNSpecifications
PRDX2 | |
Polyclonal | |
Unconjugated | |
ELISA (1:16000) Western Blot (0.3-1 ug/mL) The optimal working dilution should be determined by the end user. | |
PRDX2 | |
PRDX2 | |
A synthetic peptide corresponding to human PRDX2. | |
100 μg | |
Primary | |
This antibody is expected to recognize isoforms a and b only (NP_00580.03 and NP_859427.1 respectively). | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Western Blot | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of PRDX2. | |
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
MGC4104/NKEFB/PRP/PRX2/PRXII/TDPX1/TSA | |
Goat | |
Antigen affinity purification | |
RUO | |
7001 | |
Human, Mouse | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only