Learn More
Prealbumin, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against Prealbumin.
Manufacturer:  Abnova Corporation PAB14025
Catalog No. 89-137-806
Description
Description
Specifications
Specifications
Prealbumin
Unconjugated
The optimal working dilution should be determined by the end user.
Liquid
Purified human Prealbumin.
RUO
Primary
Recognizes human Prealbumin. Cross-reacts with mouse Prealbumin in immunoblot.
Western Blot
Goat polyclonal antibody raised against Prealbumin.
In 10mM sodium phosphate buffer, 150mM NaCl, pH 7.2 (0.05% sodium azide)
Goat
1 mg
Store at 4°C.
Polyclonal
Human, Murine